Perfluoro-2-bromo-2-methylpentane 97%
Lieferant: Apollo Scientific
PC1374LG-25G
APOSPC1374LG-25GEA
458
EUR
APOSPC1374LG-25G
Perfluoro-2-bromo-2-methylpentane 97%
Perfluoro-2-bromo-2-methylpentane
Formel:
C₆BrF₁₃ Siedepunkt: 99…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077551 CAS-Nummer: 22528-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...