2-Fluor-3-nitrobenzoesäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC1387-100G
PC1387-25G
PC1387-5G
APOSPC1387-100GEA
136
EUR
APOSPC1387-100G
APOSPC1387-25G
APOSPC1387-5G
2-Fluor-3-nitrobenzoesäure 99%
2-Fluoro-3-nitrobenzoic acid
Formel:
C₇H₄FNO₄ Molecular Weight: 185.11 g/mol Schmelzpunkt: 137…139 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797825 CAS-Nummer: 317-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...