1-Brom-3,5-difluor-2-iodbenzol 97%

Lieferant: Apollo Scientific
Achtung

PC1392-100G
APOSPC1392-100GEA 108 EUR
APOSPC1392-100G
1-Brom-3,5-difluor-2-iodbenzol 97%
1-Bromo-3,5-difluoro-2-iodobenzene
Formel: C₆H₂BrF₂I
Molecular Weight: 318.89 g/mol
Siedepunkt: 103…104 °C/15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00042183
CAS-Nummer: 175278-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR