4'-Brom-2'-fluoracetanilid
Lieferant: Apollo Scientific
Synonyme:
N-(4-Brom-2-fluorphenyl)acetamid
PC1395-100G
APOSPC1395-100GEA
234
EUR
APOSPC1395-100G
4'-Brom-2'-fluoracetanilid
4'-Brom-2'-fluoracetanilid
Formel:
H₃CC(O)NH(C₆H₃BrF) Molecular Weight: 232,05 g/mol Schmelzpunkt: 142…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042588 CAS-Nummer: 326-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...