2-Chlor-3-fluor-5-methylpyridin
Lieferant: Apollo Scientific
|
Achtung
|
PC1401-1G
APOSPC1401-1GEA
21.9
EUR
APOSPC1401-1G
2-Chlor-3-fluor-5-methylpyridin
2-Chloro-3-fluoro-5-methylpyridine
Formel:
C₆H₅ClFN Molecular Weight: 145,56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658238 CAS-Nummer: 34552-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...