1-[2-(Trifluoromethoxy)benzyl]piperazine
Lieferant: Apollo Scientific
|
Achtung
|
PC1411-1G
PC1411-25G
PC1411-5G
APOSPC1411-1GEA
0
EUR
APOSPC1411-1G
APOSPC1411-25G
APOSPC1411-5G
1-[2-(Trifluoromethoxy)benzyl]piperazine
1-[2-(Trifluoromethoxy)benzyl]piperazine
Formel:
C₁₂H₁₅F₃N₂O Siedepunkt: 78…80 °C (0,02 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368743 CAS-Nummer: 886762-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...