5-Brom-2-fluorbenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1420M-100G
APOSPC1420M-100GEA
71.2
EUR
APOSPC1420M-100G
5-Brom-2-fluorbenzonitril 98%
5-Brom-2-fluorbenzonitril
Formel:
C₇H₃BrFN Molecular Weight: 200.01 g/mol Siedepunkt: 255 Schmelzpunkt: 74…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143424 CAS-Nummer: 179897-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...