5-Brom-2-fluorbenzylamin Hydrochlorid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromo-2-fluorophenyl)methanamine hydrochloride

PC1423GD-1G PC1423GD-25G PC1423GD-5G
APOSPC1423GD-1GEA 67.2 EUR
APOSPC1423GD-1G APOSPC1423GD-25G APOSPC1423GD-5G
5-Brom-2-fluorbenzylamin Hydrochlorid 97%
5-Brom-2-fluorbenzylamin Hydrochlorid
Formel: C₇H₈BrClFN
Schmelzpunkt: 221…226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143426
CAS-Nummer: 202865-69-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR