5-Brom-2-fluorzimtsäure 98%

Lieferant: Apollo Scientific

Synonyme: 5-Brom-2-fluorcinnamsäure, 3-(5-Brom-2-fluorphenyl)propensäure

PC1423T-25G
APOSPC1423T-25GEA 65.4 EUR
APOSPC1423T-25G
5-Brom-2-fluorzimtsäure 98%
5-Brom-2-fluorzimtsäure
Formel: C₉H₆BrFO₂
Molecular Weight: 245,05 g/mol
Schmelzpunkt: 194…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143427
CAS-Nummer: 202865-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR