4-(Difluoromethoxy)benzeneboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-(Difluoromethoxy)phenyl)boronic acid
PC1431-1G
APOSPC1431-1GEA
94.4
EUR
APOSPC1431-1G
4-(Difluoromethoxy)benzeneboronic acid
4-(Difluoromethoxy)benzeneboronic acid
Formel:
C₇H₇BF₂O₃ Molecular Weight: 187,936 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09864667 CAS-Nummer: 688810-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...