1-(4-Fluorobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

PC1454-1G
APOSPC1454-1GEA 441 EUR
APOSPC1454-1G
1-(4-Fluorobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid 95%
1-(4-Fluorobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid
Formel: C₁₁H₁₀FN₃O₂
Schmelzpunkt: 203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797392
CAS-Nummer: 885950-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR