3-Fluoro[1,1'-biphenyl]-4-carbonitrile 95%

Lieferant: Apollo Scientific
Achtung

PC1456-250MG
APOSPC1456-250MGEA 21.9 EUR
APOSPC1456-250MG
3-Fluoro[1,1'-biphenyl]-4-carbonitrile 95%
3-Fluoro[1,1'-biphenyl]-4-carbonitrile
Formel: C₁₃H₈FN
Molecular Weight: 197,21 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797374
CAS-Nummer: 503177-15-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR