5-Fluor-2-methoxybenzonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluor-2-methoxybenzoesäurenitril

PC14564-1G PC14564-25G PC14564-5G
APOSPC14564-1GEA 21.9 EUR
APOSPC14564-1G APOSPC14564-25G APOSPC14564-5G
5-Fluor-2-methoxybenzonitril
5-Fluor-2-methoxybenzonitril
Formel: C₈H₆FNO
Molecular Weight: 151.14 g/mol
Schmelzpunkt: 117…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683503
CAS-Nummer: 189628-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR