4-[5-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC1461-1G
PC1461-250MG
PC1461-5G
APOSPC1461-1GEA
423
EUR
APOSPC1461-1G
APOSPC1461-250MG
APOSPC1461-5G
4-[5-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde 95%
4-[5-(Trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
Formel:
C₁₃H₈F₃NO Schmelzpunkt: 92…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797372 CAS-Nummer: 871252-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...