6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC1486-1G
APOSPC1486-1GEA
49.5
EUR
APOSPC1486-1G
6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline 95%
6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
Formel:
C₁₁H₆BrClF₃N Schmelzpunkt: 118…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153079 CAS-Nummer: 123637-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...