(4-Chloro-3-nitrophenyl)(4-fluorophenyl)methanone 97%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-4'-fluoro-3-nitrobenzophenone

PC1491-5G
APOSPC1491-5GEA 95.7 EUR
APOSPC1491-5G
(4-Chloro-3-nitrophenyl)(4-fluorophenyl)methanone 97%
(4-Chloro-3-nitrophenyl)(4-fluorophenyl)methanone
Formel: C₁₃H₇ClFNO₃
Molecular Weight: 279,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840494
CAS-Nummer: 31431-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR