2-Brom-4-(trifluormethyl)anisol 98%

Lieferant: Apollo Scientific
Achtung

PC1494-25G PC1494-5G
APOSPC1494-25GEA 81.2 EUR
APOSPC1494-25G APOSPC1494-5G
2-Brom-4-(trifluormethyl)anisol 98%
2-Brom-4-(trifluormethyl)anisol
Formel: C₈H₆BrF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD03789249
CAS-Nummer: 402-10-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR