1,3-Dichloro-2,5-difluorobenzene 95+%

Lieferant: Apollo Scientific

PC1498-1G
APOSPC1498-1GEA 127 EUR
APOSPC1498-1G
1,3-Dichloro-2,5-difluorobenzene 95+%
1,3-Dichloro-2,5-difluorobenzene
Formel: C₆H₂Cl₂F₂
Molecular Weight: 182,983 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658263
CAS-Nummer: 2367-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR