3-Chloro-2,4-difluorobenzonitrile 98%

Lieferant: Apollo Scientific

PC1503-1G
APOSPC1503-1GEA 108 EUR
APOSPC1503-1G
3-Chloro-2,4-difluorobenzonitrile 98%
3-Chloro-2,4-difluorobenzonitrile
Formel: C7H2ClF2N
Molecular Weight: 173,5500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658267
CAS-Nummer: 887267-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR