2,3-Difluoro-6-(trifluoromethyl)benzonitrile 95+%

Lieferant: Apollo Scientific
Achtung

PC1513-1G
APOSPC1513-1GEA 154 EUR
APOSPC1513-1G
2,3-Difluoro-6-(trifluoromethyl)benzonitrile 95+%
2,3-Difluoro-6-(trifluoromethyl)benzonitrile
Formel: C₈H₂F₅N
Lagertemperatur: Kühlschrank
MDL: MFCD06658270
CAS-Nummer: 186517-05-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR