4-Brom-2,6-difluorbenzonitril
Lieferant: Apollo Scientific
|
Gefahr
|
PC1515-25G
PC1515-5G
PC1515-100G
APOSPC1515-25GEA
87
EUR
APOSPC1515-25G
APOSPC1515-5G
APOSPC1515-100G
4-Brom-2,6-difluorbenzonitril
4-Brom-2,6-difluorbenzonitril
Formel:
C₇H₂BrF₂N Schmelzpunkt: 79…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310981 CAS-Nummer: 123843-67-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...