N-(4-Fluorophenyl)-1,3,5-triazine-2,4-diamine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1526-250MG
APOSPC1526-250MGEA
58
EUR
APOSPC1526-250MG
N-(4-Fluorophenyl)-1,3,5-triazine-2,4-diamine 97%
N-(4-Fluorophenyl)-1,3,5-triazine-2,4-diamine
Formel:
C₉H₈FN₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01320741 CAS-Nummer: 1549-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...