3,4-Dichlor-2-nitro-6-(trifluormethyl)toluol 97%

Lieferant: Apollo Scientific

Synonyme: 4,5-Dichloro-2-methyl-3-nitrobenzotrifluoride

PC1541-25G
APOSPC1541-25GEA 118 EUR
APOSPC1541-25G
3,4-Dichlor-2-nitro-6-(trifluormethyl)toluol 97%
3,4-Dichlor-2-nitro-6-(trifluormethyl)toluol
Formel: C₈H₄Cl₂F₃NO₂
Molecular Weight: 274,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972788
CAS-Nummer: 115571-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR