Methyl-4-(trifluormethylthio)benzoat
Lieferant: Apollo Scientific
|
Achtung
|
PC1546-1G
PC1546-25G
PC1546-5G
APOSPC1546-1GEA
0
EUR
APOSPC1546-1G
APOSPC1546-25G
APOSPC1546-5G
Methyl-4-(trifluormethylthio)benzoat
Methyl-4-(trifluormethylthio)benzoat
Formel:
C₉H₇F₃O₂S Siedepunkt: 117 °C/20mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203725 CAS-Nummer: 88489-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...