2-Brom-5-(trifluormethoxy)phenol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Brom-2-hydroxy-4-(trifluormethoxy)benzol

PC1571-25G PC1571-5G
APOSPC1571-25GEA 125 EUR
APOSPC1571-25G APOSPC1571-5G
2-Brom-5-(trifluormethoxy)phenol 98%
2-Brom-5-(trifluormethoxy)phenol
Formel: C₇H₄BrF₃O₂
Molecular Weight: 257.01 g/mol
Dichte: 1.74 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD06660162
CAS-Nummer: 205371-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR