2-Methoxy-4-(trifluoromethoxy)bromobenzene 96%

Lieferant: Apollo Scientific

PC1574-10G PC1574-1G PC1574-5G PC1574-25G
APOSPC1574-10GEA 492 EUR
APOSPC1574-10G APOSPC1574-1G APOSPC1574-5G APOSPC1574-25G
2-Methoxy-4-(trifluoromethoxy)bromobenzene 96%
2-Methoxy-4-(trifluoromethoxy)bromobenzene
Formel: C₈H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08458014
CAS-Nummer: 848360-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR