2-Fluorophenylacetyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
PC1581-25G
PC1581-5G
APOSPC1581-25GEA
274
EUR
APOSPC1581-25G
APOSPC1581-5G
2-Fluorophenylacetyl chloride
2-Fluorophenylacetyl chloride
Formel:
C₈H₆ClFO Siedepunkt: 203…204 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840709 CAS-Nummer: 451-81-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...