3-Fluorophenylacetyl chloride 97+%

Lieferant: Apollo Scientific
Gefahr

PC1582-25G PC1582-5G PC1582-100G
APOSPC1582-25GEA 0 EUR
APOSPC1582-25G APOSPC1582-5G APOSPC1582-100G
3-Fluorophenylacetyl chloride 97+%
3-Fluorophenylacetyl chloride
Formel: C₈H₆ClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD07787556
CAS-Nummer: 458-04-8
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR