Nε-Trifluoracetyl-L-lysin 98%

Lieferant: Apollo Scientific

Synonyme: H-Lys(Tfa)-OH, ε-TFA-lysine, (S)-2-Amino-6-(2,2,2-trifluoroacetamido)hexanoic acid

PC1591-100G PC1591-500G
APOSPC1591-100GEA 64.2 EUR
APOSPC1591-100G APOSPC1591-500G
Nε-Trifluoracetyl-L-lysin 98%
Nε-Trifluoracetyl-L-lysin
Formel: C₈H₁₃F₃N₂O₃
Molecular Weight: 242.20 g/mol
Schmelzpunkt: 258 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037223
CAS-Nummer: 10009-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR