2-Bromo-1,1,2-trifluoroethyl ethyl ether 97%

Lieferant: Apollo Scientific
Achtung

PC1592-1G
APOSPC1592-1GEA 40.6 EUR
APOSPC1592-1G
2-Bromo-1,1,2-trifluoroethyl ethyl ether 97%
2-Bromo-1,1,2-trifluoroethyl ethyl ether
Formel: C₄H₆BrF₃O
Siedepunkt: 116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042465
CAS-Nummer: 380-78-9
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR