2'-Bromo-4'-(trifluoromethyl)acetanilide 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1593Y-10G
PC1593Y-1G
PC1593Y-5G
APOSPC1593Y-10GEA
92.8
EUR
APOSPC1593Y-10G
APOSPC1593Y-1G
APOSPC1593Y-5G
2'-Bromo-4'-(trifluoromethyl)acetanilide 97%
2'-Bromo-4'-(trifluoromethyl)acetanilide
Formel:
C₉H₇BrF₃NO Schmelzpunkt: 144…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067782 CAS-Nummer: 175135-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...