4'-Brom-3'-(trifluormethyl)acetanilid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1594-1G
PC1594-5G
APOSPC1594-1GEA
21.9
EUR
APOSPC1594-1G
APOSPC1594-5G
4'-Brom-3'-(trifluormethyl)acetanilid 98%
4'-Brom-3'-(trifluormethyl)acetanilid
Formel:
C₉H₇BrF₃NO Schmelzpunkt: 116…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052361 CAS-Nummer: 41513-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...