4-(Difluoromethyl)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

PC1598-1G PC1598-250MG PC1598-5G
APOSPC1598-1GEA 0 EUR
APOSPC1598-1G APOSPC1598-250MG APOSPC1598-5G
4-(Difluoromethyl)benzeneboronic acid
4-(Difluoromethyl)benzeneboronic acid
Formel: C₇H₇BF₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD09056705
CAS-Nummer: 946525-43-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR