4-(Difluoromethyl)benzeneboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC1598-1G
PC1598-250MG
PC1598-5G
APOSPC1598-1GEA
0
EUR
APOSPC1598-1G
APOSPC1598-250MG
APOSPC1598-5G
4-(Difluoromethyl)benzeneboronic acid
4-(Difluoromethyl)benzeneboronic acid
Formel:
C₇H₇BF₂O₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD09056705 CAS-Nummer: 946525-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...