4-Fluoro-2-hydroxybenzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

PC1626-1G
APOSPC1626-1GEA 79.9 EUR
APOSPC1626-1G
4-Fluoro-2-hydroxybenzeneboronic acid 97%
4-Fluoro-2-hydroxybenzeneboronic acid
Formel: C₆H₆BFO₃
Schmelzpunkt: 176…180 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115662
CAS-Nummer: 850568-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR