2-Chlor-6-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlor-6-fluorbenzolboronsäure, Dihydroxy(2-chlor-6-fluorphenyl)boran, (2-Chlor-6-fluorphenyl)metaborsäure

PC1633-25G
APOSPC1633-25GEA 134 EUR
APOSPC1633-25G
2-Chlor-6-fluorphenylboronsäure 98%
2-Chlor-6-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174.37 g/mol
Schmelzpunkt: 131…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039892
CAS-Nummer: 313545-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR