3-(Difluoromethyl)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(Difluoromethyl)phenyl)boronic acid

PC1641-1G
APOSPC1641-1GEA 109 EUR
APOSPC1641-1G
3-(Difluoromethyl)benzeneboronic acid
3-(Difluoromethyl)benzeneboronic acid
Formel: C₇H₇BF₂O₂
Molecular Weight: 171.937 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09864670
CAS-Nummer: 854690-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR