5-Fluor-(2-methoxycarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2-methoxycarboxyphenylboronic acid, 5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid, 2-Methoxycarbonyl-5-fluorophenylboronic acid

PC1662-1G PC1662-5G
APOSPC1662-1GEA 128 EUR
APOSPC1662-1G APOSPC1662-5G
5-Fluor-(2-methoxycarbonyl)phenylboronsäure 98%
5-Fluor-(2-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
Molecular Weight: 197.96 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03095367
CAS-Nummer: 850568-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR