5-Fluor-(2-methoxycarbonyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluoro-2-methoxycarboxyphenylboronic acid, 5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid, 2-Methoxycarbonyl-5-fluorophenylboronic acid
PC1662-1G
PC1662-5G
APOSPC1662-1GEA
128
EUR
APOSPC1662-1G
APOSPC1662-5G
5-Fluor-(2-methoxycarbonyl)phenylboronsäure 98%
5-Fluor-(2-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BFO₄ Molecular Weight: 197.96 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03095367 CAS-Nummer: 850568-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...