3',5'-Bis(trifluormethyl)-2-chloracetanilid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(Chloroacetyl)-3,5-bis(trifluoromethyl)aniline, N-(3,5-Bis(trifluoromethyl)phenyl)-2-chloroacetamide

PC1687H-1G PC1687H-5G PC1687H-250MG
APOSPC1687H-1GEA 29 EUR
APOSPC1687H-1G APOSPC1687H-5G APOSPC1687H-250MG
3',5'-Bis(trifluormethyl)-2-chloracetanilid 98%
3',5'-Bis(trifluormethyl)-2-chloracetanilid
Formel: C₁₀H₆ClF₆NO
Molecular Weight: 305.6042 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077474
CAS-Nummer: 790-75-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR