2-Chloro-N-(3-fluorophenyl)acetamide 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(Chloroacetyl)-3-fluoroaniline
PC1688E-1G
APOSPC1688E-1GEA
21.9
EUR
APOSPC1688E-1G
2-Chloro-N-(3-fluorophenyl)acetamide 95%
2-Chloro-N-(3-fluorophenyl)acetamide
Formel:
C₈H₇ClFNO Molecular Weight: 187,6 g/mol Schmelzpunkt: 121…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01034343 CAS-Nummer: 350-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...