2-Chloro-N-[4-(trifluoromethoxy)phenyl]acetamide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(Chloroacetyl)-4-(trifluoromethoxy)aniline

PC1688T-1G
APOSPC1688T-1GEA 23.2 EUR
APOSPC1688T-1G
2-Chloro-N-[4-(trifluoromethoxy)phenyl]acetamide 97%
2-Chloro-N-[4-(trifluoromethoxy)phenyl]acetamide
Formel: C₉H₇ClF₃NO₂
Molecular Weight: 253,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00219796
CAS-Nummer: 161290-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR