2-Chloro-2'-(trifluoromethyl)acetanilide 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1689-1G
APOSPC1689-1GEA
30.6
EUR
APOSPC1689-1G
2-Chloro-2'-(trifluoromethyl)acetanilide 97%
2-Chloro-2'-(trifluoromethyl)acetanilide
Formel:
C₉H₇ClF₃NO Schmelzpunkt: 92…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00018893 CAS-Nummer: 3792-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...