Kalium(4-methylphenyl)trifluoroborat 96%
Lieferant: Apollo Scientific
|
Achtung
|
PC1697-1G
PC1697-25G
PC1697-5G
APOSPC1697-1GEA
0
EUR
APOSPC1697-1G
APOSPC1697-25G
APOSPC1697-5G
Kalium(4-methylphenyl)trifluoroborat 96%
Kalium(4-methylphenyl)trifluoroborat
Formel:
C₇H₇BF₃K Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318171 CAS-Nummer: 216434-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...