Kalium(2,4-dichlorphenyl)trifluoroborat 96%

Lieferant: Apollo Scientific

PC1701-1G PC1701-25G PC1701-5G
APOSPC1701-1GEA 0 EUR
APOSPC1701-1G APOSPC1701-25G APOSPC1701-5G
Kalium(2,4-dichlorphenyl)trifluoroborat 96%
Kalium(2,4-dichlorphenyl)trifluoroborat
Formel: C₆H₃BCl₂F₃K
Schmelzpunkt: 98…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115745
CAS-Nummer: 192863-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR