Kalium(4-isobutylphenyl)trifluoroborat 96%

Lieferant: Apollo Scientific

PC1705-1G PC1705-25G PC1705-5G
APOSPC1705-1GEA 152 EUR
APOSPC1705-1G APOSPC1705-25G APOSPC1705-5G
Kalium(4-isobutylphenyl)trifluoroborat 96%
Kalium(4-isobutylphenyl)trifluoroborat
Formel: C₁₀H₁₃BF₃K
Schmelzpunkt: 290…298 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115761
CAS-Nummer: 850623-66-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR