Kalium(3-(methoxycarbonyl)-5-nitrophenyl)trifluoroborat 96%
Lieferant: Apollo Scientific
PC1709-1G
PC1709-25G
PC1709-5G
APOSPC1709-1GEA
93.1
EUR
APOSPC1709-1G
APOSPC1709-25G
APOSPC1709-5G
Kalium(3-(methoxycarbonyl)-5-nitrophenyl)trifluoroborat 96%
Kalium(3-(methoxycarbonyl)-5-nitrophenyl)trifluoroborat
Formel:
C₈H₆BF₃KNO₄ Schmelzpunkt: 280…284 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115749 CAS-Nummer: 850623-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...