Kalium(2-(benzyloxy)phenyl)trifluoroborat 95%

Lieferant: Apollo Scientific

PC1711-10G PC1711-1G PC1711-5G
APOSPC1711-10GEA 698 EUR
APOSPC1711-10G APOSPC1711-1G APOSPC1711-5G
Kalium(2-(benzyloxy)phenyl)trifluoroborat 95%
Kalium(2-(benzyloxy)phenyl)trifluoroborat
Formel: C₁₃H₁₁BF₃KO
Schmelzpunkt: 200…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115742
CAS-Nummer: 850623-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR