3-Chlor-2,6-dibrom-4-fluoranilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dibromo-3-chloro-4-fluoroaniline, 1-Amino-3-chlor-2,6-dibrom-4-fluorbenzol
PC1717-100G
PC1717-25G
PC1717-5G
APOSPC1717-100GEA
623
EUR
APOSPC1717-100G
APOSPC1717-25G
APOSPC1717-5G
3-Chlor-2,6-dibrom-4-fluoranilin 97%
3-Chlor-2,6-dibrom-4-fluoranilin
Formel:
C₆H₃Br₂ClFN Schmelzpunkt: 97…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042199 CAS-Nummer: 175135-09-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...