Kalium(2-formylphenyl)trifluoroborat 96%

Lieferant: Apollo Scientific

PC1722-1G PC1722-5G
APOSPC1722-1GEA 67.2 EUR
APOSPC1722-1G APOSPC1722-5G
Kalium(2-formylphenyl)trifluoroborat 96%
Kalium(2-formylphenyl)trifluoroborat
Formel: C₇H₅BF₃KO
Schmelzpunkt: 220…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093979
CAS-Nummer: 192863-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR